Skip to Content
Merck
All Photos(1)

Key Documents

32939

Supelco

Zearalenone

reference material

Synonym(s):

F-2 toxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O5
CAS Number:
Molecular Weight:
318.36
Beilstein:
1350216
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

C[C@H]1CCCC(=O)CCC\C=C\c2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/b7-3+/t12-/m0/s1

InChI key

MBMQEIFVQACCCH-QBODLPLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zearalenone is one of the estrogenic metabolites produced by Fusarium species. It is usually found to co-occur with trichothecenes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Zearalenone has been used as an analytical reference standard for the determination of the analyte in:
  • Beer samples by liquid chromatography-mass spectrometry (LC-MS).

It may be used as an analytical reference standard for the determination of the analyte in:
  • Cereals by gas chromatography-mass spectrometry (GC-MS).
  • Maize samples by high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS).

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct determination of the estrogenic compounds 8-prenylnaringenin, zearalenone, ?- and ?-zearalenol in beer by liquid chromatography-mass spectrometry.
Maragou NC, et al.
Journal of Chromatography A, 1202(1), 47-57 (2008)
Cristina Juan et al.
Food chemistry, 134(4), 2389-2397 (2013-02-28)
Although analytical methods have been already reported for legislated mycotoxins as trichothecenes and zearalenone (ZON) separately, we describe the optimization of a simple and rapid multimycotoxin method for the determination of a total of 12 mycotoxins simultaneously, nine trichothecenes (NIV
Thomas J Turbyville et al.
Journal of natural products, 69(2), 178-184 (2006-02-28)
In an effort to discover small molecule inhibitors of Hsp90, we have screened over 500 EtOAc extracts of Sonoran desert plant-associated fungi using a two-stage strategy consisting of a primary cell-based heat shock induction assay (HSIA) followed by a secondary
S De Baere et al.
Analytica chimica acta, 756, 37-48 (2012-11-28)
A sensitive and specific method for the quantitative determination of zearalenone (ZEN) and its major metabolites (α-zearalenol (α-ZEL), β-zearalenol (β-ZEL), α-zearalanol (α-ZAL), β-zearalanol (β-ZAL) and zearalanone (ZAN)) in animal plasma using liquid chromatography combined with heated electrospray ionization (h-ESI) tandem
Abdellah Zinedine et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45(1), 1-18 (2006-10-19)
Zearalenone (ZEA) is a mycotoxin produced mainly by fungi belonging to the genus Fusarium in foods and feeds. It is frequently implicated in reproductive disorders of farm animals and occasionally in hyperoestrogenic syndromes in humans. There is evidence that ZEA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service