Skip to Content
Merck
All Photos(1)

Key Documents

31569

Supelco

4-Nitroaniline

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
Beilstein:
508690
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

260 °C/100 mmHg (lit.)

mp

146-149 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H,7H2

InChI key

TYMLOMAKGOJONV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitroaniline is an aromatic amine, used as an intermediate in the production of p-phenylenediamine, azo dyes, antioxidants, fuel additives, corrosion inhibitors and pesticides.

Application

4-Nitroaniline may be used as an analytical standard for the determination of 4-nitrophenol in water samples by competitive flow immunoassay coupled with a fluorescence detector.
Product of chromogenic reactions.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

415.4 °F - closed cup

Flash Point(C)

213.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A kinetic study on the degradation of p-nitroaniline by Fenton oxidation process.
Sun JH, et al.
Journal of Hazardous Materials, 148(1-2), 172-177 (2007)
Amit Kumar Mandal et al.
Scientific reports, 10(1), 5286-5286 (2020-03-27)
Cellular and tissue imaging in the second near-infrared window (NIR-II, ~1000-1350 nm) is advantageous for in vivo studies because of low light extinction by biological constituents at these wavelengths. However, deep tissue imaging at the single molecule sensitivity has not been
Competitive flow immunoassay with fluorescence detection for determination of 4-nitrophenol.
Nistor C, et al.
Analytica Chimica Acta, 426(2), 185-195 (2001)
Krzysztof Iwaszczuk et al.
Optics express, 17(24), 21969-21976 (2009-12-10)
We present a new method for data acquisition in time-resolved terahertz spectroscopy experiments. Our approach is based on simultaneous collection of reference and differential THz scans. Both the optical THz generation beam and the pump beam are modulated at two
Kunquan Li et al.
Journal of hazardous materials, 178(1-3), 553-559 (2010-03-06)
Activated carbon fibers (ACFs) were prepared for the removal of p-nitroaniline (PNA) from cotton stalk by chemical activation with NH(4)H(2)PO(4) and subsequent physical activation with steam. Surface properties of the prepared ACFs were performed using nitrogen adsorption, FTIR spectroscopy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service