Skip to Content
Merck
All Photos(1)

Key Documents

19542

Supelco

Procyanidin B1

analytical standard

Synonym(s):

(−)-Epicatechin (4β-8)-(+)-catechin, cis,trans′′-4,8′′-Bi-(3,3′,4′,5,7-Pentahydroxyflavane), Proanthocyanidin B1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H26O12
CAS Number:
Molecular Weight:
578.52
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4cc(O)cc(O)c34)c5ccc(O)c(O)c5)c2O[C@@H]1c6ccc(O)c(O)c6

InChI

1S/C30H26O12/c31-13-7-20(37)24-23(8-13)41-29(12-2-4-16(33)19(36)6-12)27(40)26(24)25-21(38)10-17(34)14-9-22(39)28(42-30(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,22,26-29,31-40H,9H2/t22-,26+,27+,28+,29+/m0/s1

InChI key

XFZJEEAOWLFHDH-UKWJTHFESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Procyanidin B1 is a phenolic compound in fruits. It has been used widely as adulteration indicator of fruit juice. It has a strong antioxidant property.

Application

Procyanidin B1 has been used in quantifying phenolic compound using HPLC, during a study performed to understand the effect of ellagitannins, ellagic acid and volatile compounds from oak wood on procyanidin B1 content of model wines. It may have been used as reference material to determine phenolic acids from apple and pear using HPLC method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum crataegus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María-Ángeles Del-Castillo-Alonso et al.
Plant physiology and biochemistry : PPB, 109, 374-386 (2016-11-05)
In the present study we assessed the effects of ambient solar UV exclusion on leaf physiology, and leaf and berry skin phenolic composition, of a major grapevine cultivar (Tempranillo) grown under typically Mediterranean field conditions over an entire season. In
Effect of ellagitannins, ellagic acid and volatile compounds from oak wood on the (+)-catechin, procyanidin B1 and malvidin-3-glucoside content of model wines.
Jord?o, A. M., et al.
Australian Journal of Grape and Wine Research, 14.3, 260-270 (2008)
A Schieber et al.
Journal of chromatography. A, 910(2), 265-273 (2001-03-23)
A new HPLC stationary phase has been applied to the analysis of phenolic acids and flavonoids with diode array and mass spectrometric detection. The separation of 26 standard compounds was achieved within 1 h. The stationary phase displayed excellent resolution
Zala Zorenc et al.
Planta, 246(2), 217-226 (2017-03-21)
Rare red currants colors caused by low anthocyanin content in the pink and a lack of anthocyanins in the white cultivar correlated with low ANS gene expression, enzyme activity, and increased sugar/acid ratios. Changes in the contents of polyphenols, sugars
Lijun Wang et al.
PloS one, 8(5), e64664-e64664 (2013-06-07)
Proanthocyanidins (PAs) contribute to poplar defense mechanisms against biotic and abiotic stresses. Transcripts of PA biosynthetic genes accumulated rapidly in response to infection by the fungus Marssonina brunnea f.sp. multigermtubi, treatments of salicylic acid (SA) and wounding, resulting in PA

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service