Skip to Content
Merck
All Photos(3)

Documents

14007

Sigma-Aldrich

Potassium titanium oxide oxalate dihydrate

Synonym(s):

Dipotassium oxodioxalatotitanate(IV) dihydrate, Oxotitanium potassium ethanedioate hydrate (1:2:2:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4K2O9Ti · 2H2O
CAS Number:
Molecular Weight:
354.13
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: titanium
reagent type: catalyst

SMILES string

O.O.[K+].[K+].[O-]C(=O)C(=O)O[Ti](=O)OC(=O)C([O-])=O

InChI

1S/2C2H2O4.2K.2H2O.O.Ti/c2*3-1(4)2(5)6;;;;;;/h2*(H,3,4)(H,5,6);;;2*1H2;;/q;;2*+1;;;;+2/p-4

InChI key

PGGRHIGITIPOBF-UHFFFAOYSA-J

General description

Potassium titanium oxide oxalate dihydrate is used as a catalyst in organic synthesis because of its excellent water solubility. It can be easily separated from the reaction mixture after the completion of the reaction.

Application

Potassium titanium oxide oxalate (PTO) dihydrate can be used as:
  • A catalyst to synthesize chromene derivatives via three-component condensation reaction of aromatic aldehydes, malononitrile, and resorcinol/ naphthol.
  • An electrolyte to functionalize the surface of aluminum alloy with TiO2 layer by plasma electrolytic oxidation (PEO).

PTO can be used as a precursor to synthesize titanium dioxide (TiO2) nanoparticles.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sub-100? C solution processed amorphous titania nanowire thin films for high-performance perovskite solar cells.
Wu W Q, et al.
Journal of Power Sources, 329, 17-22 (2016)
Electrorheological properties of suspensions of hollow globular titanium oxide/polypyrrole particles.
Sedlacik, M.
Colloid and Polymer Science, 290(1), 41-48 (2012)
Solvent?Mediated Dimension Tuning of Semiconducting Oxide Nanostructures as Efficient Charge Extraction Thin Films for Perovskite Solar Cells with Efficiency Exceeding 16%.
Wu W Q, et al.
Advanced Energy Materials, 6(7), 1502027-1502027 (2016)
Lipei Qin et al.
Scientific reports, 8(1), 16198-16198 (2018-11-06)
Visible light-driven water splitting (VLWS) into hydrogen and oxygen is attractive and depends on efficient photocatalysts. Herein, we demonstrate the first exploration of the capability to control the morphology of nanostructured TiO2 in conjunction with the choice of a suitable
Stelios Gavrielides et al.
Faraday discussions, 215(0), 407-421 (2019-04-06)
Anthropogenic CO2 is the main contributor to the increased concentration of greenhouse gases in the atmosphere, and thus utilising waste CO2 for the production of valuable chemicals is a very appealing strategy for reducing CO2 emissions. The catalytic fixation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service