Skip to Content
Merck
All Photos(1)

Key Documents

04265

Supelco

Valeryl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxopentyl)oxy]-1-propanaminium inner salt, L-Carnitine valeryl ester, Pentanoyl-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO4
CAS Number:
Molecular Weight:
245.32
Beilstein:
5946883
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

optical activity

[α]/D -20.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤10% water

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCC)=O)CC([O-])=O

InChI

1S/C12H23NO4/c1-5-6-7-12(16)17-10(8-11(14)15)9-13(2,3)4/h10H,5-9H2,1-4H3/t10-/m1/s1

InChI key

VSNFQQXVMPSASB-SNVBAGLBSA-N

General description

Valeryl-L-carnitine belongs to the class of acylcarnitines, which are acetylated forms of L-carnitine, derived via the breakdown of amino and fatty acids.

Application

Valeryl-L-carnitine may be used as an analytical reference standard for the quantification of the analyte in biological samples and milk samples using chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acylcarnitine profiling by low-resolution LC-MS
Meierhofer D
Testing, 14(8) (2019)
Carnitine precursors and short-chain acylcarnitines in water buffalo milk
Servillo L, et al.
Journal of Agricultural and Food Chemistry, 66(30), 8142-8149 (2018)
Acylcarnitine profiling by low-resolution LC-MS
Meierhofer D
PLoS ONE, 14(8) (2019)
Matrix effect-corrected liquid chromatography/tandem mass-spectrometric method for determining acylcarnitines in human urine
Abe K, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 468(30), 187-194 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service