Skip to Content
Merck
All Photos(4)

Key Documents

EXPEXTKB

Roche

KAPA Express Extract

Synonym(s):

PCR, PCR extraction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

description

for extraction only

Quality Level

usage

sufficient for 100 reactions (KK7101)
sufficient for 250 reactions (KK7102)
sufficient for 50 reactions (KK7100)
sufficient for 500 reactions (KK7103)

shelf life

≤12 mo.

packaging

pkg of 100 U (KK7100)
pkg of 1000 U (KK7103)
pkg of 200 U (KK7101)
pkg of 500 U (KK7102)

manufacturer/tradename

Roche

technique(s)

DNA extraction: suitable
PCR: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA Express Extract is a novel thermostable protease and buffer system that allows for the extraction of polymerase chain reaction (PCR)-ready DNA in as little as 15 minutes. DNA extractions are conveniently performed in a single-tube thereby greatly reducing the risk of sample loss and contamination.
The combination of KAPA Express Extract and KAPA2G Robust HotStart® ReadyMix provides a high performance solution for rapid DNA extraction and consistent downstream amplification.

Application

KAPA Express Extract is ideally suited for the extraction of polymerase chain reaction (PCR)-ready DNA from, but not limited to, the following sample types:
  • Human tissue (formalin-fixed paraffin-embedded (FFPE) samples; blood collected in EDTA tubes or on collection cards; buccal swabs; hair follicles; forensic samples; biopsies)
  • Animal samples (ear punches; tail clippings; hair follicles; blood; bone marrow; dried or fresh tissue from mouse and other mammals)
  • Fish tissues (fin punches; fresh tissue; cold and hot smoked canned samples; ethanol-preserved samples; embryo)
  • Insects (crushed)
  • Bird feathers (calamus fragments).
  • Nosema species

Features and Benefits

Key Features:
  • Rapid extraction protocol: PCR-ready DNA in 15 minutes.
  • Versatility: single kit optimized for successful extraction from a variety of tissue types, including buccal swabs, hair follicles, fish tissue, and bird feathers.
  • Single-tube reaction minimizes risk of contamination.
  • Produces high amounts of genomic DNA of good quality.
  • DNA extracted with KAPA Express Extract is compatible with any PCR system.

Increased PCR success from formalin-fixed paraffin-embedded (FFPE) samples or tissues:
  • Rapid DNA extraction and consistent downstream amplification of challenging sample types.

Routine DNA extraction from blood sample types:
  • Efficient extraction from “Guthrie”cards, FTA Elute cards, and FTA cards.

Quick Notes:
  • Extract PCR-ready DNA from different tissue types in a quick and simple, single-tube protocol (15–20 min), without the need for hazardous chemicals or multiple washing steps.
  • Significantly reduced turnaround times and risk of sample loss and contamination.
  • Use 1 μL DNA extract per 25 μL PCR. Depending on the sample type, the extract may have to be diluted prior to PCR.
  • One extract typically yields a sufficient amount of template for 50–500 PCRs.
  • DNA extracts may be diluted in TE Buffer (1:5) or 10 mM Tris-HCl pH 8–8.5 (1:5–1:10) for long-term storage at -20°C. Extracts are stable at -20°C for ≥6 months.
  • KAPA2G Robust HotStart® ReadyMix is recommended for consistent and reliable endpoint amplification of DNA extracts generated with KAPA Express Extract.
  • No quantification of DNA required prior to PCR.

Quality

All components contained with KAPA Express Extract Kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activities, and meet strict requirements with respect to DNA contamination.

Preparation Note

Handling:
KAPA2G Robust HotStart ReadyMix may not freeze solidly, even when stored at -20°C. This will not affect the shelf-life of the product. Always ensure that the product has been fully thawed and mixed before use.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

FTA is a trademark of Qiagen Group
HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA Express Extract Enzyme 1 U/µL

  • 10X KAPA Express Extract Buffer

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristics of Nosema ceranae infection in Serbian honey bee colonies
Stevanovic J, et al.
Apidologie, 44 (2013)
Molecular Study of Some Virulence Genes in Biotype Diversity of Methicillin Resistance Staphylococcus aureus isolated from Handling Carrier and Bovine Mastitis.
Hanon B M.
International Journal of Scientific Research, 6(2), 2319-7064 (2017)
BATCH-GE: Batch analysis of Next-Generation Sequencing data for genome editing assessment
Boel A, et al.
Scientific reports (2016)
The Validity of Real Time PCR and in Situ Hybridization in the Detection of JCV in Colonic Biopsies of Patient with Colorectal Cancer
Aldrubi FI, et al.
International Journal of Current Microbiology and Applied Sciences, 4(12), 107-120 (2015)
Ivica Petrovic et al.
Frontiers in endocrinology, 11, 30-30 (2020-03-03)
Aims/Hypothesis: Galectin 3 appears to play a proinflammatory role in several inflammatory and autoimmune diseases. Also, there is evidence that galectin 3 plays a role in both type-1 and type-2 diabetes. During obesity, hematopoietic cell-derived galectin 3 induces insulin resistance.

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service