Skip to Content
Merck
All Photos(2)

Key Documents

TSTP04700

Millipore

Polycarbonate Membrane Filter, 3.0 μm Pore Size

greener alternative

Isopore, filter diam. 47 mm, hydrophilic

Synonym(s):

Isopore Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

plain filter
polycarbonate membrane
white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Isopore
Millipore

sustainability

Greener Alternative Product

parameter

0.868 L/min-cm2-psi air flow rate
140 °C max. temp.
3628 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)
~475 mL/min-cm2 water flow rate at 10 psi (typical results)

diam.

47 mm

filter diam.

47 mm

thickness

22 μm

gravimetric extractables

<1%

refractive index

n/D 1.6

matrix

Isopore

pore size

11.3 % porosity
3.0 μm pore size

bubble point

≥0.32 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Isopore membrane is a polycarbonate that allows sample visualization on membrane surface using optical or electron microscopy. Due to precise pore diameter and consistent pore size, it allows size-based sample separation with accuracy. It may be useful in chemotaxis assay, air monitoring, and epifluorescent microscopy.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Chemotaxis
  • Bioassays
  • Cytology
  • Air monitoring
  • Microplastics analysis

Isopore membrane filter has been used to:
  • filter microplastics associated with microbes from seawater samples
  • wrap and encapsulate tumor for implantation in mice to mimic anti-angiogenic (AA) therapy
  • filter out phytoplankton and large-sized particles
  • hold mammalian cell lines for ultraviolet (UV) irradiation studies

Features and Benefits

  • Isopore membranes have low background interference.
  • Particle retention on the membrane surface favorable for counting and analysis.
  • Non-hygroscopic and translucent material.
  • Rapid sample drying and analysis time.
  • Also available in a brown variety.

Caution

Possible reduction in flow rates may occur after autoclaving.

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung Won Jung et al.
Scientific reports, 11(1), 6983-6983 (2021-03-28)
We investigated the dynamics of the bacterial composition and metabolic function within Akashiwo sanguinea bloom using a 100-L indoor microcosm and metagenomic next-generation sequencing. We found that the bacterial community was classified into three groups at 54% similarity. Group I
Brittan S Scales et al.
mSphere, 6(3), e0085120-e0085120 (2021-06-10)
While it is now appreciated that the millions of tons of plastic pollution travelling through marine systems carry complex communities of microorganisms, it is still unknown to what extent these biofilm communities are specific to the plastic or selected by
Denian Wang et al.
Scientific reports, 7(1), 2365-2365 (2017-05-26)
Although anti-angiogenic (AA) therapy is widely used in clinical practice, it is often challenged by insufficient efficacy and intrinsic resistance. Some studies have reported that AA therapy can even increase tumor metastasis. However, whether this is due to a specific
Haohui Duan et al.
The Journal of cell biology, 219(6) (2020-05-12)
BRCA1/2 help maintain genomic integrity by stabilizing stalled forks. Here, we identify the E3 ligase RFWD3 as an essential modulator of stalled fork stability in BRCA2-deficient cells and show that codepletion of RFWD3 rescues fork degradation, collapse, and cell sensitivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service