Skip to Content
Merck
All Photos(1)

Key Documents

QTUMMPEIX

Milli-Q

Kit: Quantum® IX Cartridge & Millipak®

Kit with Millipak® 20 Express Filter, For use with Milli-Q® water purification systems.

Synonym(s):

Kit: Quantum® IX Cartridge & Millipak® 20 Express Final Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
NACRES:
JA.13

material

polyethersulfone filter
polypropylene and polyethylene cartridge

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Millipak®
Quantum®

pore size

0.22 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

This kit is composed of a Quantum® IX Cartridge and a Millipak® 20 Express Final Filter.
The Quantum® IX Polishing Cartridge contains a combination of purification media carefully selected to remove trace contaminants affecting specific applications.
The cartridge is constructed from polypropylene and polyethylene, which are suitable for the production of ultrapure water and are qualified for low organics and ionic extractables.
The Millipak® 0.22 μm hydrophilic membrane filter produces particulate- and bacteria-free water at the point of dispense of the water purification systems.

Application

The kit contains Organex and Ionex resins for the removal of trace levels of organics and ions.

Features and Benefits

  • Quantum® IX Polishing Cartridge has been designed to remove trace levels of organics and ions from ultrapure water
  • Millipak® 20 Express Filter is designed with a 0.22 μm PES (polyethersulfone) asymmetric membrane. The filter acts as a safe and efficient barrier against particulates and bacteria.
  • The membrane′s conical pores allow a high flow rate at a low differential pressure.
  • The membrane, housing material, and production processes have been selected to minimize organic and inorganic extractable release.
  • The membrane is supported by a rigid plastic disc with channels that ensures high water flow and resistance to pressure variations without the risk of membrane rupture.

Other Notes

Directions for Use

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: General laboratory analysis
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes. Refer to the system equipment user guide section "Using the System" (provided with the system)
  • Storage Statement: Store in a dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
QUANTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service