Skip to Content
Merck
All Photos(1)

Documents

MABE89

Sigma-Aldrich

Anti-Brm Antibody, clone 1H7A10

clone 1H7A10, from rat

Synonym(s):

SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2, ATP-dependent helicase SMARCA2, probable global transcription activator SNF2L2, sucrose nonfermenting 2-like protein 2, SNF2/SWI2-like protein 2, SWI/SNF-r

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H7A10, monoclonal

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SMARCA2(6595)

Related Categories

General description

BAF190B (BRG1-associated factor 190B), also known as protein brahma homolog (Brm), and SNF2-alpha, functions as a transcriptional coactivator. This protein contains a helicase domain and is a member of the SWI/SNF family of proteins and is part of the SWI-SNF chromatin remodeling complex. The SWI-SNF complex is involved in the activation of transcription via the remodeling of nucleosome structure in an ATP-dependent manner. This large complex contains multiple proteins. BAF190 (Brm) and Brg-1 (SNF2-beta) are the ATPase subunits of the mammalian SWI-SNF complex. Brm, Brg-1, Ini1 (integrase interactor 1, or SNF5), BAF155 (SRG3) and BAF170 are thought to comprise the functional core of the SWI-SNF complex. Other complex subunits are thought to play regulatory roles. hSNF2L and hSNF2H both appear to be homologs of Drosophila ISWI, a Brm related ATPase that is present in chromatin remodeling complexes other than SWI/SNF, including the NURF (nucleosome remodeling factor).

Specificity

Recognizes BAF190B also known as Brm

Immunogen

Recombinant protein corresponding to amino acids 48-214 of mouse Brm.

Application

Immunocytochemistry:
A representative lot of this antibody was reported to work in IC using C2C12 cells (Yasuyuki Ohkawa, SSP Stem Cell Unit, Faculty of Medicine, Kyushu University, Fukuoka, Japan.)

Immunohistochemistry:
A representative lot of this antibody was reported to work in IHC using mouse embryonic and postnatal brain (Yasuyuki Ohkawa, SSP Stem Cell Unit, Faculty of Medicine, Kyushu University, Fukuoka, Japan.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Use Anti-Brm Antibody, clone 1H7A10 (Rat Monoclonal Antibody) validated in WB, IHC to detect Brm also known as ATP-dependent helicase SMARCA2 or SNF2/SWI2-like protein 2.

Quality

Evaluated by Western Blot in K562 nuclear extract.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Brm on 10 µg of K562 nuclear extract.

Target description

~140-180 kDa observed. Uncharacterized lower molecular weight bands have been reported in some lysates and may represent splice variants and/or degradation products. See Batsché E, Yaniv M, Muchardt C. Nat Struct Mol Biol. 2006 Jan;13(1):22-29.

Physical form

Format: Purified
Protein G
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Liu et al.
Molecular medicine reports, 14(6), 5435-5442 (2016-11-15)
Isoflurane is a commonly used inhaled anesthetic, which induces apoptosis of SH‑SY5Y cells in a dose‑ and time‑dependent manner; however, the underlying mechanisms remain unknown. The authors of the present study hypothesized that a mutation in β‑amyloid precursor protein (APP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service