Skip to Content
Merck
All Photos(1)

Key Documents

MABC971

Sigma-Aldrich

Anti-Kallikrein-13/KLK13 Antibody, clone 33.1

clone 33.1, from mouse

Synonym(s):

Kallikrein-13, Kallikrein-like protein 4, KLK-L4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

33.1, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KLK13(26085)

General description

Kallikreins-related peptidases (KLKs) are proteases that are important for the desquamation process of the skin (cell shedding). Kallikreins are also associated with carcinogenesis and some serve as a novel cancer and other disease biomarkers. Kallikrein-13/KLK13 is important in lung cancer by allowing lung adenocarcinoma spreading through promoting the expression of various adhesion molecules and it in assisting in extracellular matrix protein degradation necessary for movement. Kallikrein-13/KLK13 also plays a role in salivary tumors and ovarian cancer as well as other cancers in a similar fashion. In both normal and cancerous tissues, Kallikrein-13/KLK13 is secreted into the ECM and expressed in many tissues in both the embryo and adult.

Immunogen

Recombinant protein corresponding to Human Kallikrein-13/KLK13.

Application

Immunohistochemistry Analysis: A representative lot of this antibody detected Kallikrein-13/KLK13 in Normal ovarian surface epithelium and epithelial ovarian cancer tissues (White et al., (2009) British Journal of Cancer. 101:1107-1113).

ELISA: A representative lot of this antibody detected various Kallikrein-13/KLK13 concentrations of several human tissues (Kapadia et al., (2003) Clinical chemistry. 49(1):77–86).
This Anti-Kallikrein-13/KLK13 Antibody, clone 33.1 is validated for use in Immunohistochemistry and ELISA for the detection of Kallikrein-13/KLK13.

Quality

Evaluated by Immunohistochemistry in human lung and human prostate tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Kallikrein-13/KLK13 in human prostate tissue.

Target description

30 kDa calculated

Physical form

Format: Purified
Purified mouse monoclonal IgG1k in 0.1 M Sodium Bicarbonate buffer pH8.2 without preservatives.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service