Skip to Content
Merck
All Photos(2)

Key Documents

MABC584

Sigma-Aldrich

Anti-Sulf-2 Antibody, clone 2B4

clone 2B4, from mouse

Synonym(s):

Extracellular sulfatase Sulf-2, hSulf-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
2B4, monoclonal
application:
ELISA
IHC
WB
species reactivity:
human
technique(s):
ELISA: suitable
immunohistochemistry: suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B4, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SULF2(55959)

General description

Sulf-2 is also important in neurological situations including myelination and glial cell differentiation and GDNF activity. Sulf-2 activity is also found in the kidney and important in glomerular basement membrane development and glomerular function. Finally, Sulf-2 is involved in heparan sulfate metabolism. Sulf-2 removes sulfate moieties on heparan sulfate proteoglycans which in turn modulate the heparin binding growth factor signaling activity of the cell. Sulf-2 also impacts extracellular matrix interactions and it is important during development and growth, as well as Wnt signaling and VEGF production by endothelial cells during vascularization. In disease, Sulf-2 expression in cancer is upregulated and inhibition of Sulf-2 limits cancer progression and showed increased apoptosis within tumors, thus Sulf-2 activation is being examined as a potential anti-cancer therapy.

Application

This Anti-Sulf-2 Antibody, clone 2B4 is validated for use in Western Blotting and Immunohistochemistry and ELISA for the detection of Sulf-2.
Western Blot Analysis: 1 µg/mL from a representative lot of this antibody detected Sulf-2 in transfected HEK293 cell lysates (Data courtesy of Dr. Mark Singer).

Western Blot Analysis: A representative lot of this antibody detected Sulf-2 in transfected HEK293 cell lysates, but not in non-transfected HEK293 cell lysates (Lemjabbar-Alaoui, H., et al., (2010) Oncogene 29:635–646).

Immunohistochemistry Analysis: A representative lot of this antibody detected Sulf-2 in Human lung and squamouse cell carcinoma tissue (Lemjabbar-Alaoui, H., et al., (2010) Oncogene 29:635–646).

ELISA: A representative lot of this antibody detected Sulf-2, but not Sulf-1 by ELISA (Lemjabbar-Alaoui, H., et al., (2010) Oncogene 29:635–646).

Quality

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Sulf-2 in 10 µg of A549 cell lysate.

Target description

~125 & 50 kDa observed.
Sulf-2 is synthesized as a ~125 kDa pre-protein and is subsequently cleaved into ~75 kDa and ~50 kDa fragments, which are held together by disulfide bonds in the mature protein.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service