Skip to Content
Merck
All Photos(4)

Key Documents

MABC519

Sigma-Aldrich

Anti-Trop2, clone 2G10, Azide Free Antibody

clone 2G10, from mouse

Synonym(s):

Tumor-associated calcium signal transducer 2, Cell surface glycoprotein Trop-2, Membrane component chromosome 1 surface marker 1, Pancreatic carcinoma marker protein GA733-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G10, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
inhibition assay: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TACSTD2(4070)

General description

The protein known as Trop-2, or also known as cell surface glycoprotein Trop-2, Membrane component chromosome 1 surface marker 1 or Pancreatic carcinoma marker protein GA733-1 and encoded by the gene TACSTD2/GA733-1/M1S1/TROP2, is a protein that may function as a growth factor receptor and is expressed primarily in the placenta, lung and epithelial tissues but also found in certain cancers. The protein was first identified as a carcinoma antigen by the monoclonal GA733. It is a plasma membrane bound protein that transduces an intracellular calcium signal after binding unknown ligands. Mutations in Trop2 are associated with a rare eye disease leading to blindness. Trop-2 has also recently been shown to be important in regulating fibroblast motility and lung cell proliferation via the regulation of the ERK kinase pathway as well as several critical components of the cell migration apparatus.

Immunogen

Recombinant protein corresponding to the extracellular domain of human Trop2.

Application

Immunocytochemistry Analysis: A representative lot detected Trop2 in MTE 4-14/T2 and MCF7 cells (Prof. Saverio Alberti, Oncoxx Biotec.).

Flow Cytometry Analysis: A representative lot detected Trop2 in transfected cells, Skov-3 cancer cells, and HT29 cancer cells (Prof. Saverio Alberti, Oncoxx Biotec.).

Inhibition Analysis: A representative lot inhibited tumor growth in DU145 prostate cancer cells, and in HT-29, HCT116 and Colo205 cells. (Note: When used together, Cat. No. MABC519, Anti-Trop2, clone 2G10, Azide Free and Cat. No. MABC518, Anti-Trop2, clone 2EF, Azide Free maximizes inhibition of tumor growth in vivo.) (Prof. Saverio Alberti, Oncoxx Biotec.).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Trop2 antibody, clone 2G10, Azide Free is validated for use in ICC, flow cytometry & inhibition for the detection of Trop2.

Quality

Evaluated by Immunocytochemistry Analysis in MET cells transfected with a vector expressing Trop2 or with an empty vector.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected Trop 2 in MET cells transfected with a vector expressing Trop2 and not in MET cells transfected with an empty vector.

Target description

36 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service