EZSCPCB02
Control PCB (Printed Circuit Board)
for use with EZ-Stream® Vacuum Pump.
Synonym(s):
PCB, Printed Circuit Board
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
Recommended Products
feature
repair kit
manufacturer/tradename
EZ-Stream®
application(s)
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
compatibility
for use with EZ-Stream® Vacuum Pump.
General description
Control PCB (Printed Circuit Board) is designed as a spare part for EZ-Stream® Vacuum Pump. The PCB controls the entire function of the EZ-Stream® Vacuum Pump.
Design of the system allows fast and easy replacement of this component.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.
Design of the system allows fast and easy replacement of this component.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.
Application
Control PCB (Printed Circuit Board) is used as a spare for EZ-Stream® Vacuum Pump. The PCB controls the entire function of the EZ-Stream® Vacuum Pump during the filtration process.
Components
The kit includes:
- 1 PCB.
- Applicable Screw Set.
Legal Information
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service