Skip to Content
Merck
All Photos(1)

Key Documents

CDRC60202

Milli-Q

Reverse Osmosis Cartridge

Produces RO purified water at a flow rate of 16 L/hr, For use with AFS, Elix Advantage / Essential / Reference, Milli-Q Direct / Integral and RiOs Essential systems

Synonym(s):

Reverse Osmosis Membrane – 16 L/h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

packaging

pkg of 2 units

parameter

16 L/hr flow rate

H × D

28 cm (11.7 in.) × 4.5 cm (1.8 in.)

compatibility

for use with AFS®
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Reference
for use with Milli-Q® Direct
for use with Milli-Q® Integral
for use with RiOs Essential

Looking for similar products? Visit Product Comparison Guide

General description

The Reverse Osmosis membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules (MW >200kDa)

Application

Reverse Osmosis water is used in laboratories, mainly for applications such as general rinsing, feedwater to washing machines and autoclaves, or as a feed to pure and ultrapure water purification systems.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service