Skip to Content
Merck

AP136A

Goat Anti-Rat IgG Antibody, Alkaline Phosphatase conjugate

0.6 mg/mL, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
alkaline phosphatase conjugate
Clone:
polyclonal
Application:
ELISA, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

alkaline phosphatase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

concentration

0.6 mg/mL

technique(s)

ELISA: suitable, western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Immunogen

Highly purified Rat IgG isolated from pooled normal rat serum.

Application

ELISA: 1:5,000-1:50,000

Western Blot: 1:5,000-1:50,000

Optimal working dilutions must be determined by the end user.
Goat anti-Rat IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Rat IgG for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Biochem/physiol Actions

Reacts with Rat IgG gamma chain, as well as the light chains from all rat immunoglobulin classes. Monospecific on IEP against whole rat serum. Single band on IEP at a concentration of 10 mg/mL when run against rabbit anti goat whole serum.

Physical form

RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge or vortex this product if it is not completely clear after standing for 1-2 hours at room temperature.

Preparation Note

Store lyophilized material at 2-8°C until opened. After reconstitution this product is stable for several weeks at 2-8°C as an undiluted liquid. DO NOT FREEZE. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol and store at 2-8°C for up to 12 months. Please note that the concentration of protein and buffer salts will decrease to one-half of the original after adding glycerol.

Other Notes

Replaces: AP140A

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library