Skip to Content
Merck
All Photos(2)

Key Documents

ABN87

Sigma-Aldrich

Anti-DSCR3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Down syndrome critical region protein 3, Down syndrome critical region protein A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VPS26C(10311)

General description

Trisomy of human chromosome 21 result in patients with Down′s syndrome, which is a complex developmental and neurodegenerative disease that is characterized with abnormal morphological features, hypotonia, and mental retardation. The DSCR3 (Down syndrome critical region gene 3) gene is found in this region. DSCR3 expresses Down syndrome critical region protein 3, which is reportedly expressed in many studied tissues. While it is suggested that this protein is involved in neuroexcitation and in cellular signaling, further study is still required to elucidate its role and mechanism of action.

Specificity

Other homologies: Rat and Mouse (92% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human DSCR3.

Application

Immunohistochemistry Analysis: A 1:200 dilution from a representative lot detected DSCR3 in human gastric tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-DSCR3 Antibody is validated for use in WB, IH(P) for the detection of DSCR3.

Quality

Evaluated by Western Blot in A549 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected DSCR3 in 10 µg of A549 cell lysate.

Target description

~39 kDa observed. An uncharacterized band may be observed at a molecular weight of ~160 kDa in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A549 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerrie E McNally et al.
Nature cell biology, 19(10), 1214-1225 (2017-09-12)
Following endocytosis into the endosomal network, integral membrane proteins undergo sorting for lysosomal degradation or are retrieved and recycled back to the cell surface. Here we describe the discovery of an ancient and conserved multiprotein complex that orchestrates cargo retrieval
Michael D Healy et al.
Cell, 186(10), 2219-2237 (2023-05-13)
The Commander complex is required for endosomal recycling of diverse transmembrane cargos and is mutated in Ritscher-Schinzel syndrome. It comprises two sub-assemblies: Retriever composed of VPS35L, VPS26C, and VPS29; and the CCC complex which contains twelve subunits: COMMD1-COMMD10 and the
Rebeka Butkovič et al.
Nature communications, 15(1), 7180-7180 (2024-08-22)
Commander is a multiprotein complex that orchestrates endosomal recycling of integral cargo proteins and is essential for normal development. While the structure of this complex has recently been described, how cargo proteins are selected for Commander-mediated recycling remains unclear. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service