Skip to Content
Merck
All Photos(1)

Key Documents

AB10000

Sigma-Aldrich

Anti-CHIP/STUB1 Antibody

Chemicon®, from goat

Synonym(s):

CHIP, STUB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ELISA
WB
species reactivity:
human, chimpanzee
technique(s):
ELISA: suitable
western blot: suitable
citations:
4

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, chimpanzee

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... STUB1(10273)

General description

U box domain E3 ubiquitin ligase. CHIP E3 controls both the association of Hsp70/Hsp90 chaperones with ErbB2 and the down-regulation of ErbB2 induced by inhibitors of Hsp90. CHIP-induced degradation was observed for mutant and wild-type p53, which transiently associate with molecular chaperones Hsc70 and Hsp90 and can be diverted onto a degradation pathway through this association. Also, CHIP can interact with the Smad1/Smad4 proteins and block BMP signal transduction through the ubiquitin-mediated degradation of Smad proteins.

Immunogen

Peptide with sequence DAFISENGWVEDY, from the C-terminus of the protein sequence according to NP_005852.

Application

This Anti-CHIP/STUB1 Antibody is validated for use in ELISA, WB for the detection of CHIP/STUB1.

Physical form

Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.

Analysis Note

Control
Positive Control: Highly expressed in brain, heart, skelatal muscle, pancreas and placenta. Weak expression in kidney, liver and lung.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Kanack et al.
Chembiochem : a European journal of chemical biology, 23(6), e202100633-e202100633 (2022-01-22)
The ubiquitin ligase C-terminus of Hsc70 interacting protein (CHIP) is an important regulator of proteostasis. Despite playing an important role in maintaining proteostasis, little progress has been made in developing small molecules that regulate ubiquitin transfer by CHIP. Here we
Adam J Kanack et al.
The Journal of biological chemistry, 293(8), 2735-2743 (2018-01-11)
The accumulation of misfolded proteins promotes protein aggregation and neuronal death in many neurodegenerative diseases. To counteract misfolded protein accumulation, neurons have pathways that recognize and refold or degrade aggregation-prone proteins. One U-box-containing E3 ligase, C terminus of Hsc70-interacting protein
Roberto Mota et al.
American journal of cancer research, 7(9), 1948-1958 (2017-10-06)
Cancer cachexia is a severe wasting syndrome characterized by the progressive loss of lean body mass and systemic inflammation. Up to 80% of cancer patients experience cachexia, with 20-30% of cancer-related deaths directly linked to cachexia. Despite efforts to identify
Monte S Willis et al.
Cell biochemistry and function, 31(8), 724-735 (2013-04-05)
The carboxyl terminus of Hsp70-interacting protein (CHIP) is a ubiquitin ligase/cochaperone critical for the maintenance of cardiac function. Mice lacking CHIP (CHIP-/-) suffer decreased survival, enhanced myocardial injury and increased arrhythmias compared with wild-type controls following challenge with cardiac ischaemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service