Skip to Content
Merck
All Photos(2)

Key Documents

8.24476

Sigma-Aldrich

(-)-Shikimic acid

for synthesis

Synonym(s):

(-)-Shikimic acid, (3R,4S,5R)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O5
CAS Number:
Molecular Weight:
174.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-334-2
NACRES:
NA.22

Quality Level

Assay

≥97.0% (acidimetric)

form

crystals

mp

190 °C

density

1.52 g/cm3 at 27.2 °C

storage temp.

2-30°C

SMILES string

OC1C(CC(=CC1O)C(=O)O)O

InChI

1S/C7H10O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1,4-6,8-10H,2H2,(H,11,12)

InChI key

JXOHGGNKMLTUBP-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Spec. rotation [α²0/D (c=2 in water): -185.0 - -175.0 °
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service