Skip to Content
Merck
All Photos(1)

Key Documents

8.21037

Sigma-Aldrich

Polyethylene glycol

2000, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 2000, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 2000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22

vapor pressure

<0.1 hPa

Quality Level

form

flakes

autoignition temp.

420 °C

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

4-7 (20 °C, 100 g/L in H2O)

kinematic viscosity

46-55 cSt(20 °C) (50% solution)

mp

49-52 °C

transition temp

flash point 250 °C

solubility

600 g/L

density

1.21 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

no temp limit

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 2000 is a hydrophilic linear polymer that can be used as:
  • An organic additive in the synthesis of metal nanoparticles to control the size and morphology of nanostructure.
  • A carbon source to prepare acidophilic highly fluorescent carbon dots (CDs) via the one-pot hydrothermal method.
  • A dispersion stabilizer to synthesize amorphous Fe2O3 nanoparticles by microwave irradiation method. During the synthesis, PEG-2000 inhibits nonhomogeneous precipitation to obtain homogeneous precipitation of the metal nanoparticles.

Analysis Note

Hydroxyl value: 51 - 63
Melting range (lower value): ≥ 50 °C
Melting range (upper value): ≤ 55 °C
Average molecular mass: 1900 - 2200
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of amorphous Fe2O3 nanoparticles by microwave irradiation
Liao Xuehong, et al.
Materials Letters, 50(5-6), 341-346 (2001)
Yujuan Chen et al.
ACS omega, 2(8), 5251-5259 (2017-08-31)
Acidophilic highly-photoluminescent ionic liquid (IL)-modified carbon dots (CDs) were fabricated directly from polyethylene glycol-2000 (PEG2000N) by a simple one-step hydrothermal method in a system containing an IL (1-butyl-3-methylimidazolium bromide [C4mim]Br) and hydrochloric acid (HCl). In this process, PEG2000N works as
Solvothermal synthesis and characterization of crystalline CaWO4 nanoparticles
Chen S-J, et al.
Journal of Crystal Growth, 253(1-4), 361-365 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service