8.07104
Palladium/charcoal activated
10% Pd, hydrogenation catalyst (oxidic form) for synthesis
Synonym(s):
Pd/C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352402
NACRES:
NA.22
Recommended Products
Product Name
Palladium/charcoal activated (10% Pd), hydrogenation catalyst (oxidic form) for synthesis
Quality Level
form
powder
autoignition temp.
460 °C
pH
6-7 (20 °C, 50 g/L in H2O, slurry)
bulk density
400 kg/m3
storage temp.
2-30°C
Application
- Combining the use of solid-supported transition metal catalysis with microwave irradiation in solution-phase parallel library synthesis.: This research explores the combination of solid-supported transition metal catalysis, including palladium on activated charcoal, with microwave irradiation to enhance the efficiency of solution-phase parallel library synthesis. This method allows for the rapid synthesis of diverse chemical libraries, making it particularly useful for drug discovery and development (Organ MG et al., 2003).
Analysis Note
Assay (ICP-OES): 9.0 - 11.0 %
Suitable for hydrogenation catalyst: passes test
Identity (ICP-OES): passes test
Suitable for hydrogenation catalyst: passes test
Identity (ICP-OES): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service