8.01602
1-Bromobutane
for synthesis
Synonym(s):
1-Bromobutane, Butyl bromide
About This Item
vapor pressure
52 hPa ( 20 °C)
Quality Level
Assay
≥98.0% (GC)
form
liquid
autoignition temp.
265 °C
potency
2761 mg/kg LD50, oral (Rat)
expl. lim.
2.6-6.6 % (v/v)
mp
-112 °C
transition temp
flash point 18.3 °C
solubility
0.6 g/L
density
1.27 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
BrCCCC
InChI
1S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3
InChI key
MPPPKRYCTPRNTB-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.271 - 1.276
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Liver, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
50.0 °F - closed cup
Flash Point(C)
10 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service