Skip to Content
Merck
All Photos(1)

Key Documents

8.00990

Sigma-Aldrich

2-Ethyl-1-hexanol

for synthesis

Synonym(s):

2-Ethyl-1-hexanol, Isocotyl alcohol, Isooctanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH2CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-234-3
NACRES:
NA.22
Assay:
≥99% (GC)
bp:
184 °C/1013 hPa
vapor pressure:
0.13 hPa ( 20 °C)

vapor pressure

0.13 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

270-330 °C (DIN 51794)

potency

3730 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rat)

expl. lim.

1.1-12.7 % (v/v)

pH

7 (20 °C, 1 g/L in H2O)

bp

184 °C/1013 hPa

mp

-76 °C

transition temp

flash point 75 °C (DIN 51758)

density

0.832 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCC(CCCC)CC

InChI

1S/C8H18O/c1-3-5-6-8(4-2)7-9/h8-9H,3-7H2,1-2H3

InChI key

YIWUKEYIRIRTPP-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 0.832 - 0.833
Water (K. F.): ≤ 0.2 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service