Skip to Content
Merck
All Photos(1)

Documents

662045

Sigma-Aldrich

Anti-Uncoupling Protein-1 (UCP-1) (145-159) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... UCP1(7350)

General description

Protein A purified rabbit polyclonal antibody. Recognizes the UCP-1 protein.
Recognizes the UCP-1 protein.
This Anti-Uncoupling Protein-1 (UCP-1) (145-159) Rabbit pAb is validated for use in Immunoblotting for the detection of Uncoupling Protein-1 (UCP-1) (145-159).

Immunogen

Mouse
a synthetic peptide [(C)SHLHGIKPRYTGTYN] corresponding to amino acids 145-159 of mouse UCP1

Application

Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Does not exhibit cross-reactivity with UCP2 or UCP3. Since this antigen sequence is highly conserved, cross-reactivity across a broad range of species is expected. Variables associated with assay conditions will dictate the proper working dilution.
Kozak, L.P., et al. 1988. J. Biol. Chem. 263, 12274.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Traci A Czyzyk et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(4), 1151-1159 (2009-11-18)
General opioid receptor antagonists reduce food intake and body weight in rodents, but the contributions of specific receptor subtypes are unknown. We examined whether genetic deletion of the kappa-opioid receptor (KOR) in mice alters metabolic physiology. KOR-knockout (KO) and wild-type
Marcella Funicello et al.
PloS one, 2(8), e683-e683 (2007-08-02)
Growing evidences indicate that proteases are implicated in adipogenesis and in the onset of obesity. We previously reported that the cysteine protease cathepsin K (ctsk) is overexpressed in the white adipose tissue (WAT) of obese individuals. We herein characterized the
C L Plaisier et al.
Nutrition & diabetes, 2, e46-e46 (2013-03-01)
A better understanding of the processes influencing energy expenditure could provide new therapeutic strategies for reducing obesity. As the metabolic activity of the brown adipose tissue (BAT) and skeletal muscle is an important determinant of overall energy expenditure and adiposity
Laurent Vergnes et al.
The Journal of clinical endocrinology and metabolism, 101(11), 4440-4448 (2016-11-05)
Patients with pheochromocytoma (pheo) show presence of multilocular adipocytes that express uncoupling protein 1 within periadrenal (pADR) and omental (OME) fat depots. It has been hypothesized that this is due to adrenergic stimulation by catecholamines produced by the pheo tumors.
Traci A Czyzyk et al.
American journal of physiology. Endocrinology and metabolism, 305(2), E282-E292 (2013-05-30)
Obesity continues to be a global health problem, and thus it is imperative that new pathways regulating energy balance be identified. Recently, it was reported: (Hayashi K, Cao T, Passmore H, Jourdan-Le Saux C, Fogelgren B, Khan S, Hornstra I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service