Skip to Content
Merck
All Photos(1)

Key Documents

400087

Sigma-Aldrich

HIF-2α Translation Inhibitor

The HIF-2α Translation Inhibitor, also referenced under CAS 882268-69-1, controls the biological activity of HIF-2α. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

HIF-2α Translation Inhibitor, Hypoxia-Inducible Factor-2α Translation Inhibitor, Methyl-3-(2-(cyano(methylsulfonyl)methylene)hydrazino)thiophene-2-carboxylate, HIF Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O4S2
CAS Number:
Molecular Weight:
287.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (sum of two isomers)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

amber

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thienylhydrazone compound that suppresses the translation of cellular HIF-2α message by enhancing the binding of IRP1 (Iron-Regulatory Protein 1) to the IRE (Iron-Responsive Element) region at the 5′ UTR of the HIF-2α mRNA, exhibiting otherwise no effects toward the transcription of HIF-2α mRNA or the stability of HIF-2α mRNA and protein. Shown to reduce cellular protein levels of HIF-2α and HIF-2α target genes, including Glut-1, IGFBP3, and VEGF, in 786-O cells (5 to 10 µM final concentration) that express only the 2α isoform of hypoxia-inducible factor.
A cell-permeable thienylhydrazone compound that suppresses the translation of cellular HIF-2α message by enhancing the binding of IRP1 (Iron-Regulatory Protein 1) to the IRE (Iron-Responsive Element) region at the 5′ UTR of the HIF-2α mRNA, exhibiting otherwise no effects toward the transcription of HIF-2α mRNA or the stability of HIF-2α mRNA and protein. Shown to reduce cellular protein levels of HIF-2α and HIF-2α target genes, including Glut-1, IGFBP3, and VEGF, in 786-O cells (5-10 µM final concentration) that express only the 2α isoform of hypoxia-inducible factor.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zimmer, M., et al. 2008. Mol. Cell32, 838.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mai M Alzaydi et al.
American journal of respiratory cell and molecular biology, 68(1), 103-115 (2022-10-21)
Mitochondrial fission and a metabolic switch from oxidative phosphorylation to glycolysis are key features of vascular pathology in pulmonary arterial hypertension (PAH) and are associated with exuberant endothelial proliferation and apoptosis. The underlying mechanisms are poorly understood. We describe the
Lijiao Zhang et al.
International journal of chronic obstructive pulmonary disease, 17, 2383-2399 (2022-10-04)
Skeletal muscle dysfunction is an important comorbidity in patients with chronic obstructive pulmonary disease (COPD), and is associated with poor quality of life and reduced survival, but the mechanisms involved remain elusive. Ferroptosis is a newly discovered type of cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service