Skip to Content
Merck
All Photos(1)

Key Documents

382120

Sigma-Aldrich

Histone Acetyltransferase Activator, CTB

The Histone Acetyltransferase Activator, CTB, also referenced under CAS 451491-47-7, modulates the biological activity of Histone. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Acetyltransferase Activator, CTB, EP300 Activator, E1A binding protein 300 Activator, N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-ethoxybenzamide, N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-ethoxybenzamide, EP300 Activator, E1A binding protein 300 Activator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClF3NO2
CAS Number:
Molecular Weight:
343.73
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A benzamide compound that activates the histone acetyltransferase (HAT) activity of p300/EP300/E1A binding protein 300 in a dose-dependent manner (~6-fold of basal HAT activity of recombinant human p300 toward purified HeLa core histones in the presence of 250 µM CTB). SERS spectrum indicates that CTB interacts with p300 preferentially via the amide groups of the α-helix and β-sheets, resulting in altered enzyme conformation, which presumably accounts for the increased enzymatic activity.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mantelingu, K., et al. 2007. J. Phys. Chem. B111, 4527.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service