Skip to Content
Merck
All Photos(1)

Key Documents

234101

Sigma-Aldrich

Coenzyme A trilithium salt

≥94% (UV), solid, cofactor in enzymatic acetyl transfer reactions, Calbiochem®

Synonym(s):

Coenzyme A, Trilithium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H33N7O16P3S · 3Li
CAS Number:
Molecular Weight:
785.33
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

product name

Coenzyme A, Trilithium Salt, Coenzyme A, Trilithium Salt, CAS 18439-24-2, serves as a cofactor in enzymatic acetyl transfer reactions.

Quality Level

Assay

≥94% (UV)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C21H36N7O16P3S.3Li/c1-21(2,16(31)19(32)24-4-3-12(29)23-5-6-48)8-41-47(38,39)44-46(36,37)40-7-11-15(43-45(33,34)35)14(30)20(42-11)28-10-27-13-17(22)25-9-26-18(13)28;;;/h9-11,14-16,20,30-31,48H,3-8H2,1-2H3,(H,23,29)(H,24,32)(H,36,37)(H,38,39)(H2,22,25,26)(H2,33,34,35);;;/q;3*+1/p-3/t11-,14-,15-,16+,20-;;;/m1.../s1

InChI key

QSCBPHBAFBVXRK-FFKZSOHASA-K

Application

Coenzyme A, Trilithium Salt - CAS 18439-24-2- Calbiochem has been used to study the effects on CoA pathway T-cell anti-tumor immunity in mouse models

Biochem/physiol Actions

Coenzyme A (CoA) is an essential cofactor in living organisms and plays a crucial role in carboxylic acid metabolism, which includes short and long-chain fatty acids. It is a derivative of pantothenic acid. CoA participates in the transport of the acyl group and activates carbonyl groups in the tricarboxylic acid cycle and fatty acid metabolism. Over a hundred different reactions utilize CoA as it is the source of 4′-phosphopantetheine, which acts as a prosthetic group of carrier proteins of polyketide, fatty acid, and non-ribosomal peptide synthases

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael St Paul et al.
Cell metabolism, 33(12), 2415-2427 (2021-12-09)
Metabolic programming is intricately linked to the anti-tumor properties of T cells. To study the metabolic pathways associated with increased anti-tumor T cell function, we utilized a metabolomics approach to characterize three different CD8+ T cell subsets with varying degrees of anti-tumor activity
Roberta Leonardi et al.
Progress in lipid research, 44(2-3), 125-153 (2005-05-17)
Coenzyme A (CoA) is a ubiquitous essential cofactor that plays a central role in the metabolism of carboxylic acids, including short- and long-chain fatty acids. In the last few years, all of the genes encoding the CoA biosynthetic enzymes have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service