1.20571
PharmPrep® P100 RP-18e, 10 μm Scout Column
L × diam. 250 mm × 4.6 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
spherical silica gel
Quality Level
description
L × diam. 250 mm × 4.6 mm
product line
PharmPrep®
parameter
≤40 bar max. pressure
L × diam.
250 mm × 4.6 mm
matrix active group
Spherical Silica Gel
pore size
10 nm mean pore size
application(s)
plasma purification
plasma purification
separation technique
reversed phase
General description
Spherical porous Silica with uniform and homogeneous gel matrix with excellent batch to batch reproducibility
Features and Benefits
PharmPrep® P100 RP-8e, 10 µm Scout Column provides users with :
- Uniform and homogenous silica gel matrix with excellent batch-to-batch reproducibility over many column packings
- Narrow particle size distribution for high performance and high productivity in small peptide purification processes
- Reproducible specific surface area and pore size distribution
- Low back pressure
- Enhanced mechanical stability for long sorbent life-time
- a perfect separation of insulin and the main impurities desamido-insulin.
Packaging
Plastic bottle
Legal Information
PHARMPREP is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service