1.07661
GranuCult® Lactose broth
FDA (BAM), for Salmonella spp., For the performance and confirmation of the presumptive test to detect coliform
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
Recommended Products
Agency
APHA
EP
USP
Quality Level
reg. compliance
FDA (BAM)
form
medium granules (dehydrated (DCM))
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
pH
6.7-7.1 (25 °C, 13 g/L in H2O)
bulk density
650 kg/m3
application(s)
food and beverages
microbiology
storage temp.
15-25°C
suitability
Salmonella spp.
Related Categories
General description
Lactose Broth is an inhibitor-free granulated medium for the use in the presumptive testing for coliforms, especially E. coli, and the enrichment of Salmonella. The granules ensure rapid and uniform dissolution in water, it also prevents clumping of the medium and inhalation of the airborne powder. Lactose utilization is indicated by gas production. The gas liberated is collected in Durham′s tubes. Peptone present in the medium supply nitrogenous and carbonaceous compounds, long-chain amino acids, and other essential nutrients to the organisms. Lactose is a fermentable carbohydrate for the coliforms.
Application
Lactose Broth is recommended for the performance and confirmation of the presumptive test to detect coliform bacteria in food, water, dairy, and Pharmaceutical products.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service