1.07177
n-Pentane
for analysis EMSURE®
Synonym(s):
Pentane
About This Item
vapor density
2.48 (vs air)
Quality Level
product line
EMSURE®
Assay
≥99% (GC)
form
liquid
autoignition temp.
500 °F
potency
>2000 mg/kg LD50, oral (Rat)
expl. lim.
~8.3 %
impurities
≤0.000002% B (Boron)
≤0.00005% Al (Aluminium)
≤0.0002 meq/g Acidity
≤0.005% Sulfur compounds (as S)
≤0.01% Water
evapn. residue
≤0.001%
color
APHA: ≤10
refractive index
n20/D 1.358 (lit.)
bp
35-36 °C (lit.)
mp
−130 °C (lit.)
transition temp
flash point -48 °C (refers to pure substance)
solubility
0.4 g/L
density
0.626 g/mL at 25 °C (lit.)
cation traces
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
SMILES string
CCCCC
InChI
1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3
InChI key
OFBQJSOFQDEBGM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Catalytic Cracking to Light Olefins: n-Pentane is extensively used in studies focused on catalytic cracking processes. Recent research has explored the use of extra-framework aluminum in ZSM-5 zeolites to enhance the production of light olefins from n-pentane. This application is crucial for improving the efficiency of catalytic processes and the production of valuable petrochemical feedstocks (He et al., Journal of Analytical and Applied Pyrolysis, 2022).
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-40.0 °F
Flash Point(C)
-40 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service