1.06585
di-Sodium hydrogen phosphate
anhydrous, EMPROVE® EXPERT, Ph. Eur., BP, JPE, USP, E 339
Pharma Manufacturing
Synonym(s):
Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate
About This Item
Recommended Products
Agency
BP
JPE
Ph. Eur.
USP
Quality Level
product line
EMPROVE® EXPERT
form
solid
pH
8.7-9.3 (20 °C, 10 g/L in H2O)
pKa (25 °C)
(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)
mp
250 °C (decomposition)
application(s)
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
2-30°C
SMILES string
[Na+].[Na+].[H]OP([O-])([O-])=O
InChI
1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2
InChI key
BNIILDVGGAEEIG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
also commonly purchased with this product
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Method development for titer determination of three different monoclonal antibodies - cetuximab, trastuzumab, and universal antibody standard using a Chromolith® WP 300 Protein A column.
Method development for titer determination of three different monoclonal antibodies - cetuximab, trastuzumab, and universal antibody standard using a Chromolith® WP 300 Protein A column.
Method development for titer determination of three different monoclonal antibodies - cetuximab, trastuzumab, and universal antibody standard using a Chromolith® WP 300 Protein A column.
Method development for titer determination of three different monoclonal antibodies - cetuximab, trastuzumab, and universal antibody standard using a Chromolith® WP 300 Protein A column.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service