Skip to Content
Merck
All Photos(2)

Key Documents

1.00662

SAFC

Sorbic acid

EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, NF, FCC, E 200

Pharma Manufacturing

Synonym(s):

Sorbic acid, 2,4-Hexadienic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352106
E Number:
E200
EC Index Number:
203-768-7

Agency

BP
ChP
NF
Ph. Eur.

Quality Level

reg. compliance

FCC

vapor pressure

0.00018 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

99.0-101.0% substance (anhydrous) basis (acidimetric)

form

solid

autoignition temp.

>130 °C

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

pH

3.3 (20 °C, 1.6 g/L in H2O)

bp

160-260 °C/1013 hPa (decomposition)

mp

120-150 °C

solubility

1.6 g/L

density

1.2 g/cm3 at 20 °C

cation traces

As: ≤0.0003%
Cd: ≤0.001%
Cu: ≤0.001%
Hg: ≤0.0001%
Pb: ≤0.0002%
Zn: ≤0.001%
heavy metals (as Pb): ≤0.0010%

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

OC(=O)\C=C\C=C\C

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sorbic acid is commonly used as a preservative in pharmaceutical formulations and cosmetics. It is also used as additive for the preservation of food.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service