Skip to Content
Merck
All Photos(1)

Key Documents

07-758

Sigma-Aldrich

Anti-phospho-Flt-1 (Tyr1213) Antibody

Upstate®, from rabbit

Synonym(s):

Vascular endothelial growth factor receptor 1 (VEGFR-1), Vascular permeability factor receptor, Tyrosine-protein kinase FRT, Fms-like tyrosine kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, human, goat, sheep, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1213)

Gene Information

bovine ... Flt1(503620)
human ... FLT1(2321)
mouse ... Flt1(14254)
rat ... Flt1(54251)

Specificity

Recognizes phospho-Flt-1 (Tyr1213).

Immunogen

KLH-conjugated, synthetic peptide containing the sequence (...VR[pY]VN...) in which [pY] corresponds to phospho-tyrosine at residue 1213 of human Flt-1. The immunizing sequence is identical in marmoset, has 14/15 identical amino acids in rat, goat, sheep, cow, and rhesus monkey and 13/15 identical amino acids in mouse.

Application

Anti-phospho-Flt-1 (Tyr1213) Antibody detects level of phospho-Flt-1 (Tyr1213) & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Routinely evaluated by immunoblot on recombinant Flt-1 (Catalog # 14-562) phosphorylated upon incubation with ATP

Target description

~180 kDa (66 kDa for the fragment tested in QC)

Physical form

ImmunoAffinity Purified
Immunoaffinity purified rabbit IgG in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Recombinant Flt-1 (Catalogue No. 14-562) phosphorylated by ATP

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuanke Zhao et al.
Cancer science, 106(11), 1596-1606 (2015-08-25)
Angiogenesis is a prerequisite of tumor growth and metastasis and, thus, anti-angiogenesis treatment has become an important part of cancer therapy. A 15-amino acid peptide of the fibrinogen α chain, fibrinostatin, was previously found in serum samples of gastric cancer
Zheng Zhou et al.
American journal of cancer research, 5(10), 3149-3161 (2015-12-23)
Angiogenesis is central to the growth of cancers and VEGFR-1/Flt-1 plays an important role during the neovascularization under pathological conditions. We previously founded a VEGFR1 antagonistic peptide, F56, by screening the phage peptide library. We showed that DHFR-F56 chimeric protein
Liuying Hu et al.
Journal of dermatological science (2022-10-10)
Although vitamins or their derivatives (Vits), such as panthenyl ethyl ether, tocopherol acetate, and pyridoxine, have been widely used in topical hair care products, their efficacy and mode of action have been insufficiently studied. To elucidate the biological influence of
EGFR- and VEGF(R)-targeted small molecules show synergistic activity in colorectal cancer models refractory to combinations of monoclonal antibodies.
Poindessous, V; Ouaret, D; El Ouadrani, K; Battistella, A; Megalophonos, VF; Kamsu-Kom et al.
Clinical cancer research : an official journal of the American Association for Cancer Research null
Paul Mésange et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(11), 2548-2558 (2018-03-02)
Purpose: There is extensive cross-talk between VEGF- and EGFR-pathway signaling in colorectal cancer. However, combinations of VEGF- and EGFR-targeted monoclonal antibodies (mAb) show disappointing activity, in particular for patients with mutant RAS Previous results show that tyrosine kinase inhibitors (TKI)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service