Skip to Content
Merck
All Photos(2)

Key Documents

857123P

Avanti

16:0 Lyso PA

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-sn-glycero-3-phosphate (sodium salt); palmitoyl lysophosphatidic acid; PA(16:0/0:0); 16:0 LPA; 110678

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O7PNa
CAS Number:
Molecular Weight:
432.46
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-palmitoyl-2-hydroxy-sn-glycero-3-phosphate (sodium salt)

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer, TLC)

form

powder

packaging

pkg of 1 × 200 mg (857123P-200mg)
pkg of 1 × 25 mg (857123P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidic acid (LPA) belongs to a class of phospholipids, that has a glycerol backbone attached by a phosphate group, an aliphatic chain and a hydroxyl group. It has a phosphate group attached to sn-3 position, an aliphatic chain attached to sn-1 or sn-2 position and the hydroxyl group attached to the remaining sn-1 or sn-2 position.

Application

16:0 Lyso PA is suitable to use as an internal standard to extract lipids from plasma sample of ovarian cancer patients. It is also suitable for the preparation of lipid solutions for synthesizing liposomes.

Biochem/physiol Actions

Lysophosphatidic acid (LPA) participates in several biological functions. It acts as a potential biomarker for detecting ovarian cancer. LPA stimulates platelet aggregation, smooth muscle contraction, chemotaxis, cytoskeleton rearrangement, Ca2+ mobilization, neurotransmitter release and cell proliferation. Palmitoyl-lysophosphatidic acid helps in increasing aggregation, stimulated by adenosine diphosphate (ADP).

Packaging

5 mL Amber Glass Screw Cap Vial (857123P-200mg)
5 mL Amber Glass Screw Cap Vial (857123P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The SH3 domain of Caskin1 binds to lysophosphatidic acid suggesting a direct role for the lipid in intracellular signaling
Koprivanacz K, et al.
Cellular Signalling, 32(1-2), 66-75 (2017)
Maya Sawada et al.
Langmuir : the ACS journal of surfaces and colloids, 36(17), 4671-4681 (2020-04-10)
Amyloid fibrils are formed by denatured proteins when the supersaturation of denatured proteins is broken by agitation, such as ultrasonication, or by seeding, although the detailed mechanism of how solubility and supersaturation regulate amyloid formation remains unclear. To further understand
Are lysophosphatidic acids or phosphatidic acids involved in stimulus activation coupling in platelets
Benton AM, et al.
Blood, 60(3), 642-649 (1982)
Quantitative determination of lysophosphatidic acid by LC/ESI/MS/MS employing a reversed phase HPLC column
Shan L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 864(1-2), 22-28 (2008)
Qingxin Shi et al.
Food & function, 10(12), 7782-7792 (2019-11-30)
Triterpenoid saponins from Kuding tea have demonstrated preventive effects on hyperlipidaemia induced by a high-fat diet. Lysoglycerophospholipids (Lyso-GPLs) are known to be associated with proatherogenic conditions such as hyperlipidaemia. In this study, a target profiling strategy based on a multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service