Skip to Content
Merck
All Photos(2)

Key Documents

840497P

Avanti

16:0-18:2 PG

1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

1-hexadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(16:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H74O10PNa
CAS Number:
Molecular Weight:
768.97
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840497P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840497P

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H75O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,37-38,41-42H,3-10,12,14-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b13-11-,18-17-;/t37?,38-;/m1./s1

InChI key

JBOGSUDWCXWRFD-YBBFZHODSA-M

General description

16:0-18:2 PG or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of palmitic acid, linolenic acid and phospho-rac-(1-glycerol) at the sn-1, sn-2 and sn-3 positions respectively, in the glycerol backbone. Phosphatidylglycerol (PG) is an anionic lipid, enriched in several bacterial membranes. It is present in animal tissues, mainly in the lung surfactant.

Application

16:0-18:2 PG or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) might be used as a phospholipid standard to identify phospholipid classes in phosphorus assay. It might also be used in liposomes to study its effect on the proliferation of mouse keratinocytes, in vitro.

Biochem/physiol Actions

Phosphatidylglycerol (PG) is useful in preparing nanoliposomes.

Packaging

5 mL Clear Glass Sealed Ampule (840497P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 117(5), 223-249 (2008)
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 117(5), 223-249 (2008)
Pulmonary Surfactant Trafficking and Homeostasis
Lung Epithelial Biology in the Pathogenesis of Pulmonary Disease, 117(5), 59-75 (2017)
Nanoparticles: A novel approach to target tumors
Nano- and Microscale Drug Delivery Systems, 117(5), 113-129 (2017)
Anna M Russano et al.
The Journal of allergy and clinical immunology, 117(5), 1178-1184 (2006-05-06)
Evidences from mice and human beings indicate that gammadelta T cells could be relevant in recognition of stress-induced self and/or yet unidentified inhaled foreign antigens. Their specificity differs from classic MHC-restricted alphabeta T cells and involves the immunoglobulin-like structure of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service