Skip to Content
Merck
All Photos(3)

Key Documents

840027P

Avanti

E. coli PE

Avanti Research - A Croda Brand 840027P, powder

Synonym(s):

L-α-phosphatidylethanolamine (E. coli)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840027P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840027P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[NH3+])(OC[C@@]([H])(COC(CCCCCCCCCCCCCCC)=O)OC(CCCCCCC/C=C/CCCCCCCC)=O)=O

General description

L-α-phosphatidylethanolamine (E. coli) is the major phospholipid constituting around 70-80%, present in the inner membrane of E. coli. PE in E. coli tends to form non-bilayer structure.

Application

E. coli PE may be used to study the physical states and thermodynamic properties extracted from E. coli.

Biochem/physiol Actions

L-α-phosphatidylethanolamine (E. coli) plays a vital role in cytoskeletal organization during cytokinesis. It also interacts with several membrane proteins such as lactose permease.

Packaging

5 mL Clear Glass Sealed Ampule (840027P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Depletion of phosphatidylethanolamine affects secretion of Escherichia coli alkaline phosphatase and its transcriptional expression
Mikhaleva N, et al.
Febs Letters, 493(2-3), 85-90 (2001)
Phosphatidylethanolamine-phosphatidylglycerol bilayer as a model of the inner bacterial membrane
Murzyn K, et al.
Biophysical Journal, 88(2), 1091-1103 (2005)
Physical states and thermodynamic properties of model gram-negative bacterial inner membranes
Hoyo J, et al.
Chemistry and Physics of Lipids, 218, 57-64 (2019)
K Emoto et al.
The Journal of cell biology, 149(6), 1215-1224 (2000-06-13)
Phosphatidylethanolamine (PE) is a major membrane phospholipid that is mainly localized in the inner leaflet of the plasma membrane. We previously demonstrated that PE was exposed on the cell surface of the cleavage furrow during cytokinesis. Immobilization of cell surface
Mikhail Bogdanov et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(34), 15057-15062 (2010-08-11)
Phosphatidylcholine (PC) has been widely used in place of naturally occurring phosphatidylethanolamine (PE) in reconstitution of bacterial membrane proteins. However, PC does not support native structure or function for several reconstituted transport proteins. Lactose permease (LacY) of Escherichia coli, when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service