Skip to Content
Merck
All Photos(2)

Key Documents

840008P

Avanti

Lyso PAF (from Heart PC)

Avanti Research - A Croda Brand 840008P, powder

Synonym(s):

1-alkyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (840008P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840008P

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCCCCCCCCCC

InChI

1S/C24H52NO6P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-29-22-24(26)23-31-32(27,28)30-21-19-25(2,3)4/h24,26H,5-23H2,1-4H3/t24-/m1/s1

InChI key

VLBPIWYTPAXCFJ-XMMPIXPASA-N

General description

Lyso-platelet activating factor (lyso-PAF) is the alkyl homolog of lysophosphatidylcholine.

Application

Lyso PAF (from Heart PC) (1-alkyl-2-hydroxy-sn-glycero-3-phosphocholine) has been used to study its inhibitory actions on alkyl-PAF-induced death in Swiss albino mice. It may be used as an acyl acceptor in the determination of lysophosphatidic acid (LPA) acyltransferase (LPAAT) activity.

Biochem/physiol Actions

Lyso-platelet activating factor (lyso-PAF) can stimulate inflammation in a murine model of pleurisy. It acts as a stable metabolite and a precursor of PAF. The turnover of lyso-PAF to PAF is expected to induce continued inflammation in target organs or tissues.

Packaging

5 mL Clear Glass Sealed Ampule (840008P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Marathe et al.
Journal of lipid research, 42(9), 1430-1437 (2001-08-24)
Lysophosphatidylcholine is an abundant component of plasma and oxidized LDL that displays several biological activities, some of which may occur through the platelet-activating factor (PAF) receptor. We find that commercial lysophosphatidylcholine, its alkyl homolog (lyso-PAF), and PAF all induce inflammation
H Inoue et al.
Life sciences, 52(21), PL227-PL232 (1993-01-01)
The conversion of lyso-platelet activating factor (lyso-PAF) to PAF in cultured paranasal sinus mucosa obtained from normal human subjects was studied. The PAF concentration in the medium was determined after addition of lyso-PAF. PAF became detectable at 10 minutes after
Vyala Hanumanthareddy Chaithra et al.
Journal of lipid research, 59(11), 2063-2074 (2018-08-25)
Platelet-activating factor (PAF) is a potent inflammatory mediator that exerts its actions via the single PAF receptor (PAF-R). Cells that biosynthesize alkyl-PAF also make abundant amounts of the less potent PAF analogue acyl-PAF, which competes for PAF-R. Both PAF species
C Eberhardt et al.
The Journal of biological chemistry, 272(32), 20299-20305 (1997-08-08)
Lysophosphatidic acid (1-acyl-sn-glycero-3-phosphate (LPA)) is a phospholipid with diverse biological activities. The mediator serves as an intermediate in membrane phospholipid metabolism but is also produced in acute settings by activated platelets. LPA is converted to phosphatidic acid, itself a lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service