Skip to Content
Merck
All Photos(1)

Key Documents

810387C

Avanti

18:1 PE-TopFluor AF594

Avanti Research - A Croda Brand (810387C), chloroform solution

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-( TopFluor AF594) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H119N6O18PS2
CAS Number:
Molecular Weight:
1499.89
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 250 μg (810387C-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand (810387C)

concentration

250 μg/mL (810387C-250UG)

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (810387C-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service