Skip to Content
Merck
All Photos(1)

Key Documents

800741P

Avanti

N-18:1 L-Serine

N-oleoyl L-serine, powder

Synonym(s):

1-(9Z-octadecenoyl)-L-serine; (S)-3-hydroxy-2-(9Z)- octadecenamidopropanoate; (S)-3-hydroxy-2-oleoamidopropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H39NO4
CAS Number:
Molecular Weight:
369.54
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (800741P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800741P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

General description

N-18:1 L-Serine is a N-acyl amino acid. The amino acid and the fatty acid acyl moiety are covalently linked by amide bond.

Biochem/physiol Actions

N-acyl amino acids are signaling molecules and N-acyl-serine is an endogenous orphan lipid mediator. N-oleoyl serine mediates energy homeostasis and bone metabolism. It also participates in the mitogenic signaling in osteoblasts and is a novel antiosteoporotic drugs.

Packaging

5 mL Amber Glass Screw Cap Vial (800741P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reem Smoum et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(41), 17710-17715 (2010-09-30)
Bone mass is determined by a continuous remodeling process, whereby the mineralized matrix is being removed by osteoclasts and subsequently replaced with newly formed bone tissue produced by osteoblasts. Here we report the presence of endogenous amides of long-chain fatty
Natalia Battista et al.
Biomolecules, 9(12) (2019-12-11)
The lipid signal is becoming increasingly crowded as increasingly fatty acid amide derivatives are being identified and considered relevant therapeutic targets. The identification of N-arachidonoyl-ethanolamine as endogenous ligand of cannabinoid type-1 and type-2 receptors as well as the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service