Skip to Content
Merck
All Photos(1)

Key Documents

300115S

Avanti

Dox-NP (5mL vial)

Avanti Research - A Croda Brand

Synonym(s):

Dox-NP (5mL vial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

description

Dox-NP (Liposomal Encapsulated Doxorubicin)
frozen liposomal suspension (buffer - 10% w/w sucrose, 10 mM histidine ~pH 6.5)

Assay

>99% (HPLC/MS)

form

dispersion

packaging

pkg of 1 × 5 mL (300115S-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

General description

Formulation: A Pegylated, long-circulating liposomal formulation with doxorubicin entrapped within an ammonium sulfate core via remote-loading. All unencapsulated was removed to yield a completely liposomal doxorubicin product. Liposomes are stored frozen for a longer shelf life.

Application

Dox-NP(5mL vial) has been used as a sample to study its characteristics by asymmetrical-flow field flow fractionation (MD-AF4). It may also be used as an intravenous (i.v.) injection in flank or orthotopic animal models of cancer.

Packaging

10 mL amber serum vial (300115S-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Dox-NP is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical characterization of liposomal drug formulations using multi-detector asymmetrical-flow field flow fractionation
Parot J, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society (2020)
J Parot et al.
Journal of controlled release : official journal of the Controlled Release Society, 320, 495-510 (2020-02-01)
Liposomal formulations for the treatment of cancer and other diseases are the most common form of nanotechnology enabled pharmaceuticals (NEPs) submitted for market approval and in clinical application today. The accurate characterization of their physical-chemical properties is a key requirement;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service