Z740936
Aldrich® Essentials volumetric pipette, class A, meets DIN-ISO 648
capacity 25 mL, tol. 0.030 mL, blue
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
borosilicate glass
feature
class A
packaging
pack of 12 ea
manufacturer/tradename
Sigma-Aldrich
tol. measuring range
0.030 mL
capacity
25 mL
color
blue
Looking for similar products? Visit Product Comparison Guide
General description
Borosilicate glass, reusable, volumetric pipets, Calibrated to deliver. Permanent white fill mark and capacity mark. Color coded. Meets Class A, DIN-ISO 648 specifications.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service