Z683272
J.Young high-vacuum PTFE valve, three way
stopcock size 10 mm, side arm O.D. 13 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40141600
NACRES:
NB.77
manufacturer/tradename
J. Young
side arm O.D.
13 mm
stopcock size
10 mm
Looking for similar products? Visit Product Comparison Guide
General description
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
Features:
- PTFE piston with unique PTFE-sheathed elastomer O-ring seals
- Grease-free glass-to-PTFE seals
- High-vacuum to 10-6 torr
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service