Skip to Content
Merck
All Photos(1)

Key Documents

W349909

Sigma-Aldrich

Hexyl 2-methylbutanoate

≥95%, FCC, FG

Synonym(s):

Hexyl 2-methylbutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2(CH2)5CH3
CAS Number:
Molecular Weight:
186.29
FEMA Number:
3499
EC Number:
Council of Europe no.:
4132
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.507
NACRES:
NA.21
Organoleptic:
apple; green; spicy; fruity; waxy; tropical
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.419 (lit.)

bp

217-219 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; spicy; fruity; waxy; tropical

SMILES string

CCCCCCOC(=O)C(C)CC

InChI

1S/C11H22O2/c1-4-6-7-8-9-13-11(12)10(3)5-2/h10H,4-9H2,1-3H3

InChI key

YUECNVSODFDKOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service