Skip to Content
Merck
All Photos(2)

Key Documents

W312420

Sigma-Aldrich

Vanillylacetone

≥98%, natural (US), FG

Synonym(s):

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, Vanillyl acetone, Zingerone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CH2CH2COCH3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
3124
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.005
NACRES:
NA.21

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.510

Assay

≥98%

refractive index

n20/D 1.541 (lit.)

bp

141 °C/0.5 mmHg (lit.)

mp

40-41 °C (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

spicy; balsamic; vanilla

storage temp.

2-8°C

SMILES string

COc1cc(CCC(C)=O)ccc1O

InChI

1S/C11H14O3/c1-8(12)3-4-9-5-6-10(13)11(7-9)14-2/h5-7,13H,3-4H2,1-2H3

InChI key

OJYLAHXKWMRDGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service