Skip to Content
Merck
All Photos(1)

Key Documents

W278009

Sigma-Aldrich

trans-2,cis-6-Nonadien-1-ol

≥95%, stabilized, FCC, FG

Synonym(s):

(E,Z)-2,6-nonadien-1-ol, (E,Z)-cucumber alcohol, (E,Z)-violet leaf alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH=CHCH2CH2CH=CHCH2OH
CAS Number:
Molecular Weight:
140.22
FEMA Number:
2780
EC Number:
Council of Europe no.:
589
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.231
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥95%

may contain

~0.10% alpha-tocopherol, synthetic as stabilizer

composition

contains IFRA and EU1223/2009 restricted Benzyl Alcohol

refractive index

n20/D 1.468 (lit.)

bp

96-100 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

benzyl alcohol

Organoleptic

cucumber; green; leafy; violet

SMILES string

[H]\C(CC)=C(/[H])CC\C([H])=C(/[H])CO

InChI

1S/C9H16O/c1-2-3-4-5-6-7-8-9-10/h3-4,7-8,10H,2,5-6,9H2,1H3/b4-3-,8-7+

InChI key

AMXYRHBJZOVHOL-ODYTWBPASA-N

General description

trans-2,cis-6-nonadien-1-ol has been identified in the volatile fractions of fresh-cut honeydews and melons.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolomics in melon: a new opportunity for aroma analysis.
Allwood JW, et al.
Phytochemistry, 99, 61-72 (2014)
Characteristics of fresh-cut honeydew (Cucumis xmelo L.) available to processors in winter and summer and its quality maintenance by modified atmosphere packaging.
Bai J, et al.
Postharvest biology and technology, 28(3), 349-359 (2003)
Ping Yang et al.
Journal of agricultural and food chemistry, 67(28), 7926-7934 (2019-06-30)
Black garlic is a new garlic product produced through fermentation of fresh garlic and is very popular in Asia countries due to its health benefits. Its key aroma-active compounds were characterized by gas chromatography-olfactometry-mass spectrometry (GC-O-MS), gas chromatography-time-of-flight mass spectrometry
Xiao Yang et al.
Foods (Basel, Switzerland), 9(2) (2020-02-26)
Thermally treated watermelon juice (TW) presents a strong unpleasant smell, resulting in poor consumer acceptance. It is necessary to identify the key off-flavor compounds in TW. Solid-phase microextraction (SPME) and solvent-assisted flavor evaporation (SAFE) coupled with gas chromatography-olfactometry-mass spectrometry (GC-O-MS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service