SLW3830/04M
Pyrex® Gas distribution tubes
porosity grade 2
Synonym(s):
Pyrex, SciLabware 3830/04M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
borosilicate glass tube
colorless tube
sterility
non-sterile
feature
graduations: no
manufacturer/tradename
SciLabware 3830/04M
thimble O.D.
15 mm
tube diam.
8 mm
pore size
grade 2 porosity
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Ideal for the dispersion of gas in liquids
- Borosilicate glass tube with sintered thimble
- Choice of thimble porosities to achieve optimum gas bubble size
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service