Skip to Content
Merck
All Photos(1)

Key Documents

MT51350042

Mettler-Toledo certified buffer

pH 4.01, volume 6 x 250 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

sterility

non-sterile

form

liquid

manufacturer/tradename

Mettler-Toledo 51350042

volume

6 x 250 mL

pH

4.01

Looking for similar products? Visit Product Comparison Guide

General description

The perfect product for regulated industries. These solutions are traceable to a certificate of an accredited body of DKD.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service