Skip to Content
Merck
All Photos(1)

Key Documents

JRD0440

Sigma-Aldrich

Methyl 4-(methylthio)benzoate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2S
CAS Number:
Molecular Weight:
182.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

COC(=O)c1ccc(SC)cc1

InChI

1S/C9H10O2S/c1-11-9(10)7-3-5-8(12-2)6-4-7/h3-6H,1-2H3

InChI key

UQGNSVPCBCFZCE-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E D Kroese et al.
Mutation research, 245(2), 67-74 (1990-10-01)
Thioethers are effective scavengers of electrophilic metabolites derived from the hepatocarcinogen N-hydroxy-2-acetylaminofluorene (van den Goorbergh et al., 1987). In this study 2 of these thioethers, 4-(methylthio)benzoic acid (MTB) and its methylester, methyl 4-(methylthio)benzoate (MMTB), have been tested for their ability
J A van den Goorbergh et al.
Carcinogenesis, 8(2), 275-279 (1987-02-01)
In order to find potentially effective compounds that could prevent the covalent binding of the carcinogen N-hydroxy-2-acetylaminofluorene (N-OH-AAF) to rat liver macromolecules in vivo, the prevention of the covalent binding to RNA of the sulfate ester of the carcinogen N-OH-AAF
P S Robbins et al.
Die Naturwissenschaften, 90(11), 517-520 (2003-11-12)
The female-produced sex pheromone of Phyllophaga crinita (Burmeister) (Coleoptera: Scarabaeidae: Melolonthinae; the adult has no common name) is identified as methyl 2-(methylthio)benzoate. This is the first identification of a sulfur-containing, long-distance, female-produced sex attractant from any insect taxa. The root-feeding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service