DWK258520101
DURAN® Filter funnel
glass, flask volume (35 mL), porosity 1
Synonym(s):
258520101
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
Recommended Products
material
glass
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 258520101
disc diam.
35 mm
filter diam.
35 mm
flask volume
(35 mL)
pore size
1 porosity
capacity
50 mL capacity
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Büchner funnels are cylindrical with an integral sintered disc. Without additional filter paper, only coarse filtration is possible. Büchner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
Features and Benefits
- Manufactured from 3.3. expansion borosilicate glass
- Funnels have an integral sintered glass filtration disc
- Comprehensive range of funnel sizes and sinter porosities (grades) are available
- The use of filter paper is not required
- Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Filter funnels can be used with a filtering flask via a conical rubber seal (GUKO)
- Fired-on, highly durable white print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service