DWK216181703
Duran® Mixing Cylinder
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, joint: ST/NS 19/26, OCTAGONAL PE-STOPPER, cylinder capacity 50 mL
Synonym(s):
216181703
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
hexagonal glass cylinder (base)
Joint
joint: ST/NS19/26
feature
class B
graduated WHITE GRADUATION
packaging
pkg of 2 ea
manufacturer/tradename
DURAN® 216181703
cylinder capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
Application
Measuring liquid amounts, Diluting solutions, Mixing several components, Volumetric measurement, To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250 °C in drying cabinets or sterilisers., Boro 3.3
Features and Benefits
Calibration is based on the poured in volume (“In”) at a + 20 °C reference temperature; Large hexagonal base equipped with three knobs which increase its stability and prevent the cylinder from rolling; A uniform wall thickness over the entire measuring range prevents wedge errors; Fired-on, Easy-to-read scale with graduation marks to estimate the content; The volume content tolerances conform to accuracy Class B
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service