Skip to Content
Merck
All Photos(3)

Documents

D86604

Sigma-Aldrich

2-Diethylaminoethanethiol hydrochloride

95%

Synonym(s):

2-Diethylaminoethyl mercaptan hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCH2CH2SH · HCl
CAS Number:
Molecular Weight:
169.72
Beilstein:
3548721
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

crystals

mp

170-175 °C (lit.)

SMILES string

Cl[H].CCN(CC)CCS

InChI

1S/C6H15NS.ClH/c1-3-7(4-2)5-6-8;/h8H,3-6H2,1-2H3;1H

InChI key

QAXLTZBBDSDRPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Liu et al.
International journal of pharmaceutics, 562, 1-10 (2019-03-18)
FK506 (tacrolimus) is an effective immunosuppressant, but its poor water solubility and low bioavailability impose barriers to ocular drug delivery. The nanomicelles (NMs) formulations comprised of amino-terminated poly(ethylene glycol-block-poly(D,L)-lactic acid) (NH2-PEG-b-PLA) and hydroxypropyl methylcellulose (HPMC) were developed to increase the
Amin Jafari et al.
Materials (Basel, Switzerland), 13(4) (2020-02-23)
As a promising strategy for the treatment of various diseases, gene therapy has attracted increasing attention over the past decade. Among various gene delivery approaches, non-viral vectors made of synthetic biomaterials have shown significant potential. Due to their synthetic nature
Xu Wang et al.
Nature communications, 11(1), 5828-5828 (2020-11-19)
Stimuli-sensitive nanomaterials with cooperative response are capable of converting subtle and gradual biological variations into robust outputs to improve the precision of diagnostic or therapeutic outcomes. In this study, we report the design, synthesis and characterization of a series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service